Product Details

SNP ID
rs138279686
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:1518962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATTTACGGAAGCAGGAGGAGTC[C/T]GTGCAGAAGCAGGAAGCCATGCGGC
Phenotype
MIM: 612316 MIM: 612317
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD3A PubMed Links

Gene Details

Gene
ATAD3A
Gene Name
ATPase family, AAA domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170535.1 756 Silent Mutation TCC,TCT S162S NP_001164006.1
NM_001170536.1 756 Silent Mutation TCC,TCT S83S NP_001164007.1
NM_018188.3 756 Silent Mutation TCC,TCT S210S NP_060658.3
XM_017001628.1 756 Silent Mutation TCC,TCT S162S XP_016857117.1
Gene
ATAD3B
Gene Name
ATPase family, AAA domain containing 3B
There are no transcripts associated with this gene.

View Full Product Details