Product Details

SNP ID
rs138411002
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:154327820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGATGGATACCTTGAGAGCTGT[G/T]CCCCTTTTTTCAATATCTGGCCTCT
Phenotype
MIM: 606578 MIM: 605867
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AQP10 PubMed Links

Gene Details

Gene
AQP10
Gene Name
aquaporin 10
There are no transcripts associated with this gene.

Gene
ATP8B2
Gene Name
ATPase phospholipid transporting 8B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005855.1 21 Intron NP_001005855.1
NM_020452.3 21 Silent Mutation GTG,GTT V7V NP_065185.1
XM_005245355.2 21 Intron XP_005245412.1
XM_005245356.3 21 Intron XP_005245413.1
XM_017001871.1 21 Intron XP_016857360.1
XM_017001872.1 21 Intron XP_016857361.1

View Full Product Details