Product Details

SNP ID
rs138533259
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:155197181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTATGCCACAGGGCATTTCGGAG[A/G]TGCTCACCTGGGCCAGACACTGATG
Phenotype
MIM: 158340 MIM: 188062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR92B PubMed Links

Gene Details

Gene
MIR92B
Gene Name
microRNA 92b
There are no transcripts associated with this gene.

Gene
MUC1
Gene Name
mucin 1, cell surface associated
There are no transcripts associated with this gene.

Gene
THBS3
Gene Name
thrombospondin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252607.1 2588 Silent Mutation CAC,CAT H835H NP_001239536.1
NM_001252608.1 2588 Silent Mutation CAC,CAT H724H NP_001239537.1
NM_007112.4 2588 Silent Mutation CAC,CAT H844H NP_009043.1
XM_006711498.3 2588 Silent Mutation CAC,CAT H901H XP_006711561.1
XM_011509926.2 2588 Silent Mutation CAC,CAT H904H XP_011508228.1
XM_011509927.2 2588 Silent Mutation CAC,CAT H895H XP_011508229.1
XM_011509928.2 2588 Silent Mutation CAC,CAT H873H XP_011508230.1
XM_011509929.1 2588 Silent Mutation CAC,CAT H847H XP_011508231.1
XM_011509930.2 2588 Silent Mutation CAC,CAT H904H XP_011508232.1
XM_011509931.1 2588 Silent Mutation CAC,CAT H809H XP_011508233.1
XM_011509932.2 2588 Silent Mutation CAC,CAT H809H XP_011508234.1
XM_011509933.1 2588 Silent Mutation CAC,CAT H809H XP_011508235.1
XM_011509934.1 2588 Silent Mutation CAC,CAT H668H XP_011508236.1
XM_011509935.1 2588 Silent Mutation CAC,CAT H668H XP_011508237.1
XM_011509936.1 2588 Silent Mutation CAC,CAT H592H XP_011508238.1
XM_017002206.1 2588 Silent Mutation CAC,CAT H376H XP_016857695.1

View Full Product Details