Product Details

SNP ID
rs138610048
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:153623658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAACTTCTCAGGAAAAGGGAGAA[A/C]GTGGAAAGATGTACTGAGCTTTGAG
Phenotype
MIM: 176940 MIM: 601989 MIM: 607986
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
S100A1 PubMed Links

Gene Details

Gene
S100A1
Gene Name
S100 calcium binding protein A1
There are no transcripts associated with this gene.

Gene
S100A13
Gene Name
S100 calcium binding protein A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024210.1 Intron NP_001019381.1
NM_001024211.1 Intron NP_001019382.1
NM_001024212.1 Intron NP_001019383.1
NM_001024213.1 Intron NP_001019384.1
NM_005979.2 Intron NP_005970.1
XM_005245434.3 Intron XP_005245491.1
XM_011509862.2 Intron XP_011508164.1
XM_011509863.2 Intron XP_011508165.1
XM_011509864.1 Intron XP_011508166.1
XM_017002034.1 Intron XP_016857523.1
XM_017002035.1 Intron XP_016857524.1
XM_017002036.1 Intron XP_016857525.1
Gene
S100A14
Gene Name
S100 calcium binding protein A14
There are no transcripts associated with this gene.

View Full Product Details