Product Details

SNP ID
rs138624825
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:1523868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGAAGCACGGGTGCGCGACAT[C/T]GCCATAGCAACAAGGAACACCAAGA
Phenotype
MIM: 612316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD3A PubMed Links

Gene Details

Gene
ATAD3A
Gene Name
ATPase family, AAA domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170535.1 1263 Silent Mutation ATC,ATT I331I NP_001164006.1
NM_001170536.1 1263 Silent Mutation ATC,ATT I252I NP_001164007.1
NM_018188.3 1263 Silent Mutation ATC,ATT I379I NP_060658.3
XM_017001628.1 1263 Silent Mutation ATC,ATT I331I XP_016857117.1

View Full Product Details