Product Details

SNP ID
rs138723852
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:19256051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCGGAACGCCTGGAAGCACAGCC[A/G]GTGAGCGGGCAGGGGGAGGAAGGCC
Phenotype
MIM: 608478 MIM: 616846
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR7L PubMed Links

Gene Details

Gene
AKR7L
Gene Name
aldo-keto reductase family 7 like (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
EMC1
Gene Name
ER membrane protein complex subunit 1
There are no transcripts associated with this gene.

Gene
MRTO4
Gene Name
MRT4 homolog, ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016183.3 446 Missense Mutation CAG,CGG Q64R NP_057267.2
XM_006710675.3 446 Missense Mutation CAG,CGG Q17R XP_006710738.1

View Full Product Details