Product Details

SNP ID
rs139076791
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:230703176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCGGGTTGGCCACGCGGCCCA[A/G]GAAGTGCAGGGCAGTGGCGCTTTGA
Phenotype
MIM: 106150 MIM: 606974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGT PubMed Links

Gene Details

Gene
AGT
Gene Name
angiotensinogen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000029.3 1931 Silent Mutation CTG,TTG L475L NP_000020.1
Gene
COG2
Gene Name
component of oligomeric golgi complex 2
There are no transcripts associated with this gene.

View Full Product Details