Product Details
- SNP ID
-
rs139165394
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:16023844 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGAAGGTGTTCCGCCTGGGAGAA[G/T]ACTGGTACTTCCTGATGACCCTCGG
- Phenotype
-
MIM: 602024
MIM: 610692
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLCNKA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1537803] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLCNKA
- Gene Name
- chloride voltage-gated channel Ka
- Gene
- HSPB7
- Gene Name
- heat shock protein family B (small) member 7
There are no transcripts associated with this gene.
View Full Product Details