Product Details

SNP ID
rs139324473
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:247895873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGACAAGACCATCAGCTACATGG[A/G]CTGTGCCATCCAGCTCTTCCTGTTC
Phenotype
MIM: 616729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2W3 PubMed Links

Gene Details

Gene
OR2W3
Gene Name
olfactory receptor family 2 subfamily W member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001957.2 287 Missense Mutation GAC,GGC D96G NP_001001957.2

View Full Product Details