Product Details

SNP ID
rs139681051
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:39897443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCATCGGAGCTGTCTTTTCTCTG[C/T]AGCCATCCTCTTCTCAGCCCCCACC
Phenotype
MIM: 164850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYCL PubMed Links

Gene Details

Gene
MYCL
Gene Name
v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033081.2 1605 Missense Mutation ACA,GCA T342A NP_001028253.1
NM_001033082.2 1605 Missense Mutation ACA,GCA T372A NP_001028254.2
NM_005376.4 1605 Intron NP_005367.2

View Full Product Details