Product Details

SNP ID
rs139724628
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:162498006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCCCTTAACCCACACCGATGGC[A/G]GGATCCGGCTGCGCCTGGGGCGCGG
Phenotype
MIM: 608849
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UHMK1 PubMed Links

Gene Details

Gene
UHMK1
Gene Name
U2AF homology motif (UHM) kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184763.1 202 Intron NP_001171692.1
NM_144624.2 202 Silent Mutation GCA,GCG A2A NP_653225.2
NM_175866.4 202 Silent Mutation GCA,GCG A2A NP_787062.1

View Full Product Details