Product Details

SNP ID
rs139819219
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:175004641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAAGATCTAGAAGAGGTAAAG[G/T]TGTTGCTGGAAAAGGCTACTAGGAA
Phenotype
MIM: 606186 MIM: 611978 MIM: 609238
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CACYBP PubMed Links

Gene Details

Gene
CACYBP
Gene Name
calcyclin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007214.1 415 UTR 5 NP_001007215.1
NM_014412.2 415 Missense Mutation GTG,TTG V15L NP_055227.1
XM_017001046.1 415 UTR 5 XP_016856535.1
Gene
LOC105371622
Gene Name
uncharacterized LOC105371622
There are no transcripts associated with this gene.

Gene
MRPS14
Gene Name
mitochondrial ribosomal protein S14
There are no transcripts associated with this gene.

Gene
RABGAP1L
Gene Name
RAB GTPase activating protein 1 like
There are no transcripts associated with this gene.

View Full Product Details