Product Details

SNP ID
rs140079723
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:112674948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCAGCTGCCGGCAGCTCCGGGA[A/G]GCGGGCCAGTGTTTCGAGAGTTTCC
Phenotype
MIM: 601580 MIM: 610742
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPZA1 PubMed Links

Gene Details

Gene
CAPZA1
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
There are no transcripts associated with this gene.

Gene
MOV10
Gene Name
Mov10 RISC complex RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130079.2 535 Silent Mutation GAA,GAG E12E NP_001123551.1
NM_001286072.1 535 UTR 5 NP_001273001.1
NM_001321324.1 535 Silent Mutation GAA,GAG E12E NP_001308253.1
NM_020963.4 535 Silent Mutation GAA,GAG E12E NP_066014.1
XM_005270869.4 535 UTR 5 XP_005270926.1
XM_005270870.4 535 UTR 5 XP_005270927.1
XM_017001318.1 535 UTR 5 XP_016856807.1
XM_017001319.1 535 Silent Mutation GAA,GAG E12E XP_016856808.1

View Full Product Details