Product Details

SNP ID
rs140188244
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:1203867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACACCCACAGGTCTCCCAGCC[A/G]CCCCTTCTCCTCTGCCGATCGCTCG
Phenotype
MIM: 603905 MIM: 600315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF18 PubMed Links

Gene Details

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004195.2 820 Missense Mutation CGG,TGG R235W NP_004186.1
NM_148901.1 820 Silent Mutation GGC,GGT G164G NP_683699.1
NM_148902.1 820 Missense Mutation CGG,TGG R228W NP_683700.1
XM_017002722.1 820 Silent Mutation GGC,GGT G256G XP_016858211.1
Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
There are no transcripts associated with this gene.

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
There are no transcripts associated with this gene.

View Full Product Details