Product Details

SNP ID
rs140800433
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:145994139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCTCTCCAATCGGTGATCTTC[G/A]GGAATGACATCCATATAGCAGGGAG
Phenotype
MIM: 606599
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TXNIP PubMed Links

Gene Details

Gene
TXNIP
Gene Name
thioredoxin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313972.1 1331 Silent Mutation NP_001300901.1
NM_006472.5 1331 Silent Mutation NP_006463.3
XM_017000085.1 1331 Silent Mutation XP_016855574.1

View Full Product Details