Product Details

SNP ID
rs140981637
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:89186478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGCTGCTCCTTCTGTTTTTCT[C/T]TTAGCAGCTCCTGTTCCTTCTCAGC
Phenotype
MIM: 612466
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GBP4 PubMed Links

Gene Details

Gene
GBP4
Gene Name
guanylate binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052941.4 1678 Missense Mutation AAA,AGA K521R NP_443173.2

View Full Product Details