Product Details

SNP ID
rs141302565
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:6225199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATTTCTTCTTCTTCTGTTCGAT[C/T]GCGGAAGAATCGCCACACTGTCAGG
Phenotype
MIM: 605851 MIM: 616923
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ICMT PubMed Links

Gene Details

Gene
ICMT
Gene Name
isoprenylcysteine carboxyl methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012405.3 509 Missense Mutation AAT,GAT N246D NP_036537.1
XM_011541140.2 509 Missense Mutation AAT,GAT N150D XP_011539442.1
Gene
RNF207
Gene Name
ring finger protein 207
There are no transcripts associated with this gene.

View Full Product Details