Product Details
- SNP ID
-
rs141307739
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:155197586 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCTTGATAACTGAAGAGAAAGCC[C/T]GCGTAGTCATCATCAGTCACTGTGT
- Phenotype
-
MIM: 158340
MIM: 188062
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR92B
PubMed Links
Gene Details
- Gene
- MIR92B
- Gene Name
- microRNA 92b
There are no transcripts associated with this gene.
- Gene
- MUC1
- Gene Name
- mucin 1, cell surface associated
There are no transcripts associated with this gene.
- Gene
- THBS3
- Gene Name
- thrombospondin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001252607.1 |
2432 |
Silent Mutation |
GCA,GCG |
A783A |
NP_001239536.1 |
NM_001252608.1 |
2432 |
Silent Mutation |
GCA,GCG |
A672A |
NP_001239537.1 |
NM_007112.4 |
2432 |
Silent Mutation |
GCA,GCG |
A792A |
NP_009043.1 |
XM_006711498.3 |
2432 |
Silent Mutation |
GCA,GCG |
A849A |
XP_006711561.1 |
XM_011509926.2 |
2432 |
Silent Mutation |
GCA,GCG |
A852A |
XP_011508228.1 |
XM_011509927.2 |
2432 |
Silent Mutation |
GCA,GCG |
A843A |
XP_011508229.1 |
XM_011509928.2 |
2432 |
Silent Mutation |
GCA,GCG |
A821A |
XP_011508230.1 |
XM_011509929.1 |
2432 |
Silent Mutation |
GCA,GCG |
A795A |
XP_011508231.1 |
XM_011509930.2 |
2432 |
Silent Mutation |
GCA,GCG |
A852A |
XP_011508232.1 |
XM_011509931.1 |
2432 |
Silent Mutation |
GCA,GCG |
A757A |
XP_011508233.1 |
XM_011509932.2 |
2432 |
Silent Mutation |
GCA,GCG |
A757A |
XP_011508234.1 |
XM_011509933.1 |
2432 |
Silent Mutation |
GCA,GCG |
A757A |
XP_011508235.1 |
XM_011509934.1 |
2432 |
Silent Mutation |
GCA,GCG |
A616A |
XP_011508236.1 |
XM_011509935.1 |
2432 |
Silent Mutation |
GCA,GCG |
A616A |
XP_011508237.1 |
XM_011509936.1 |
2432 |
Silent Mutation |
GCA,GCG |
A540A |
XP_011508238.1 |
XM_017002206.1 |
2432 |
Silent Mutation |
GCA,GCG |
A324A |
XP_016857695.1 |
View Full Product Details