Product Details

SNP ID
rs141318392
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:161147189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCATCTACACTAATCTTCTATAA[C/T]GTGGCTTCCTCCCTATAGCAGAGCT
Phenotype
MIM: 610554
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UFC1 PubMed Links

Gene Details

Gene
UFC1
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016406.3 Intron NP_057490.2
XM_005245254.1 Intron XP_005245311.1
XM_017001450.1 Intron XP_016856939.1
XM_017001451.1 Intron XP_016856940.1

View Full Product Details