Product Details

SNP ID
rs141378492
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:108698672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAATGCTACGATCTTTTCTCAC[A/C]AAACTGGAGTGGTTTTCTACCTTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRPF38B PubMed Links

Gene Details

Gene
PRPF38B
Gene Name
pre-mRNA processing factor 38B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018061.2 995 Silent Mutation ACA,ACC T209T NP_060531.2
XM_011541687.2 995 UTR 3 XP_011539989.1
XM_017001596.1 995 Silent Mutation ACA,ACC T64T XP_016857085.1
XM_017001597.1 995 Silent Mutation ACA,ACC T25T XP_016857086.1
XM_017001598.1 995 Silent Mutation ACA,ACC T12T XP_016857087.1

View Full Product Details