Product Details

SNP ID
rs141394710
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:43981126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCAGGCCAGCAGCCGGATGCC[C/T]GGGCCCACTGGGCGGGCCAGTGGCC
Phenotype
MIM: 603717 MIM: 604013 MIM: 603456
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0B PubMed Links

Gene Details

Gene
ATP6V0B
Gene Name
ATPase H+ transporting V0 subunit b
There are no transcripts associated with this gene.

Gene
B4GALT2
Gene Name
beta-1,4-galactosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005417.2 129 UTR 5 NP_001005417.1
NM_003780.4 129 UTR 5 NP_003771.1
NM_030587.2 129 Missense Mutation CCG,CTG P18L NP_085076.2
XM_017002716.1 129 UTR 5 XP_016858205.1
XM_017002717.1 129 UTR 5 XP_016858206.1
Gene
DPH2
Gene Name
DPH2 homolog
There are no transcripts associated with this gene.

View Full Product Details