Product Details

SNP ID
rs142013001
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:78493177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACAAAACCAATATTTCATTCTA[C/T]GAAAATTACATCCAAACATGTGAAA
Phenotype
MIM: 600563
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PTGFR PubMed Links
Additional Information
For this assay, SNP(s) [rs12088246] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PTGFR
Gene Name
prostaglandin F receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000959.3 700 Missense Mutation ACG,ATG T145M NP_000950.1
NM_001039585.1 700 Missense Mutation ACG,ATG T145M NP_001034674.1
XM_017001873.1 700 Missense Mutation ACG,ATG T145M XP_016857362.1

View Full Product Details