Product Details

SNP ID
rs142069965
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:77882002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGGATAATAAATGTTAAAGCA[A/G]TTATATTCACAATCTGTTACAACAA
Phenotype
MIM: 616773 MIM: 613121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIGA1 PubMed Links

Gene Details

Gene
MIGA1
Gene Name
mitoguardin 1
There are no transcripts associated with this gene.

Gene
NEXN
Gene Name
nexilin F-actin binding protein
There are no transcripts associated with this gene.

Gene
NEXN-AS1
Gene Name
NEXN antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details