Product Details

SNP ID
rs142203953
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:78492829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCCAGACGGAAAACCGGCTTT[C/T]CGTATTTTTTTCAGTAATCTTCATG
Phenotype
MIM: 600563
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGFR PubMed Links

Gene Details

Gene
PTGFR
Gene Name
prostaglandin F receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000959.3 352 Missense Mutation TCC,TTC S29F NP_000950.1
NM_001039585.1 352 Missense Mutation TCC,TTC S29F NP_001034674.1
XM_017001873.1 352 Missense Mutation TCC,TTC S29F XP_016857362.1

View Full Product Details