Product Details

SNP ID
rs142215019
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:158562807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAATCTTGTTGTGGTTGAAGGT[G/T]TACATGGCCCGGGGCCGTGCATAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6P1 PubMed Links

Gene Details

Gene
OR6P1
Gene Name
olfactory receptor family 6 subfamily P member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160325.1 798 Silent Mutation NP_001153797.1

View Full Product Details