Product Details

SNP ID
rs142437742
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:155172777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGAGACCCGAACCAAGCAGGCC[A/G]CCCTGGATGGTGAGCCACTCGGTGG
Phenotype
MIM: 600986
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTCAP2 PubMed Links

Gene Details

Gene
KRTCAP2
Gene Name
keratinocyte associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173852.3 Intron NP_776251.1
Gene
TRIM46
Gene Name
tripartite motif containing 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256599.1 Intron NP_001243528.1
NM_001256600.1 Intron NP_001243529.1
NM_001256601.1 Intron NP_001243530.1
NM_001282378.1 Intron NP_001269307.1
NM_001282379.1 Intron NP_001269308.1
NM_025058.4 Intron NP_079334.3
XM_011510002.1 Intron XP_011508304.1
XM_017002393.1 Intron XP_016857882.1

View Full Product Details