Product Details

SNP ID
rs142684092
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:182056385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTCGTTGGAGGTGATTGGACCT[C/G]GTGAAGGCCTGGCCGCAGTCCTCAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF648 PubMed Links

Gene Details

Gene
ZNF648
Gene Name
zinc finger protein 648
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009992.1 5767 Silent Mutation ACC,ACG T542T NP_001009992.1
XM_017000301.1 5767 Silent Mutation ACC,ACG T542T XP_016855790.1
XM_017000302.1 5767 Silent Mutation ACC,ACG T542T XP_016855791.1

View Full Product Details