Product Details

SNP ID
rs142755741
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:10450239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCACAAGATGCCATTGTCCCCC[A/G]GCCTCCTGCTGCTGCTGCTCTCCGG
Phenotype
MIM: 609130 MIM: 602784
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APITD1 PubMed Links

Gene Details

Gene
APITD1
Gene Name
apoptosis-inducing, TAF9-like domain 1
There are no transcripts associated with this gene.

Gene
APITD1-CORT
Gene Name
APITD1-CORT readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243768.1 521 Intron NP_001230697.1
NM_001270517.1 521 Intron NP_001257446.1
NM_198544.3 521 Intron NP_940946.1
NM_199006.2 521 Intron NP_950171.2
Gene
CORT
Gene Name
cortistatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302.4 521 Missense Mutation AGC,GGC S6G NP_001293.3

View Full Product Details