Product Details

SNP ID
rs142816984
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:92231480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCACCATTATTATTAGCTCATC[C/T]CTTAAGGTAAAGGGGCATTTGGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf146 PubMed Links

Gene Details

Gene
C1orf146
Gene Name
chromosome 1 open reading frame 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012425.1 327 Silent Mutation TCC,TCT S20S NP_001012425.1
XM_011541447.2 327 Silent Mutation TCC,TCT S20S XP_011539749.1

View Full Product Details