Product Details

SNP ID
rs143233499
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:26057240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGGCTCTCAGGGCGTCTGCTAT[A/G]TGCTCTAAATCCAAAGTAAAGAAGT
Phenotype
MIM: 606131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM63 PubMed Links

Gene Details

Gene
TRIM63
Gene Name
tripartite motif containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032588.3 1082 Silent Mutation CAC,CAT H314H NP_115977.2
XM_017002559.1 1082 Silent Mutation CAC,CAT H314H XP_016858048.1

View Full Product Details