Product Details

SNP ID
rs143401010
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:6416892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAATGGGTGAAAGTAAGAGGAAC[C/G]TGCCCTCCCTCCTCTCCACACCTCC
Phenotype
MIM: 606351 MIM: 609970
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
There are no transcripts associated with this gene.

Gene
HES2
Gene Name
hes family bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019089.4 2602 UTR 3 NP_061962.2

View Full Product Details