Product Details

SNP ID
rs143402418
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:11069599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTGACTGGCTGTAGTCGTAAGG[C/T]GTAAACTCTTTTTCTGGTGGCTCTG
Phenotype
MIM: 605960 MIM: 182891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001998.2 2527 Silent Mutation ACA,ACG T816T NP_001001998.1
NM_002685.3 2527 Silent Mutation ACA,ACG T791T NP_002676.1
XM_005263475.2 2527 Silent Mutation ACA,ACG T821T XP_005263532.2
XM_017001494.1 2527 Silent Mutation ACA,ACG T405T XP_016856983.1
XM_017001495.1 2527 Silent Mutation ACA,ACG T400T XP_016856984.1
XM_017001496.1 2527 Silent Mutation ACA,ACG T389T XP_016856985.1
XM_017001497.1 2527 Silent Mutation ACA,ACG T384T XP_016856986.1
Gene
SRM
Gene Name
spermidine synthase
There are no transcripts associated with this gene.

View Full Product Details