Product Details

SNP ID
rs143514018
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:171704209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTAATATGAAATATTATAATCA[G/T]TGTTTATATACACAAATAATGAACT
Phenotype
MIM: 606909
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VAMP4 PubMed Links

Gene Details

Gene
VAMP4
Gene Name
vesicle associated membrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185127.1 1102 UTR 3 NP_001172056.1
NM_003762.4 1102 UTR 3 NP_003753.2
XM_017002713.1 1102 UTR 3 XP_016858202.1

View Full Product Details