Product Details
- SNP ID
-
rs143623408
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:41028591 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGCAGCAGCACCTACGTGTTTG[C/T]GAAACAGGTCAGCGTGGGGTCCAAG
- Phenotype
-
MIM: 616396
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCMH1
PubMed Links
Gene Details
- Gene
- SCMH1
- Gene Name
- sex comb on midleg homolog 1 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001031694.2 |
2082 |
Missense Mutation |
CAC,CGC |
H617R |
NP_001026864.1 |
NM_001172218.1 |
2082 |
Missense Mutation |
CAC,CGC |
H534R |
NP_001165689.1 |
NM_001172219.1 |
2082 |
Missense Mutation |
CAC,CGC |
H605R |
NP_001165690.1 |
NM_001172220.1 |
2082 |
Missense Mutation |
CAC,CGC |
H534R |
NP_001165691.1 |
NM_001172221.1 |
2082 |
Missense Mutation |
CAC,CGC |
H556R |
NP_001165692.1 |
NM_001172222.2 |
2082 |
Missense Mutation |
CAC,CGC |
H437R |
NP_001165693.1 |
NM_012236.3 |
2082 |
Missense Mutation |
CAC,CGC |
H548R |
NP_036368.1 |
XM_006710462.2 |
2082 |
Missense Mutation |
CAC,CGC |
H627R |
XP_006710525.1 |
XM_006710464.1 |
2082 |
Missense Mutation |
CAC,CGC |
H556R |
XP_006710527.1 |
XM_011541032.2 |
2082 |
Missense Mutation |
CAC,CGC |
H627R |
XP_011539334.1 |
XM_011541033.2 |
2082 |
Missense Mutation |
CAC,CGC |
H617R |
XP_011539335.1 |
XM_011541034.1 |
2082 |
UTR 3 |
|
|
XP_011539336.1 |
XM_011541035.2 |
2082 |
Missense Mutation |
CAC,CGC |
H570R |
XP_011539337.1 |
XM_011541036.2 |
2082 |
Missense Mutation |
CAC,CGC |
H556R |
XP_011539338.1 |
XM_011541039.1 |
2082 |
Missense Mutation |
CAC,CGC |
H507R |
XP_011539341.1 |
XM_011541040.2 |
2082 |
Intron |
|
|
XP_011539342.1 |
XM_011541043.2 |
2082 |
Missense Mutation |
CAC,CGC |
H449R |
XP_011539345.1 |
XM_011541044.1 |
2082 |
Missense Mutation |
CAC,CGC |
H449R |
XP_011539346.1 |
XM_011541045.2 |
2082 |
Missense Mutation |
CAC,CGC |
H526R |
XP_011539347.2 |
XM_017000698.1 |
2082 |
Missense Mutation |
CAC,CGC |
H575R |
XP_016856187.1 |
XM_017000699.1 |
2082 |
Missense Mutation |
CAC,CGC |
H556R |
XP_016856188.1 |
XM_017000700.1 |
2082 |
UTR 3 |
|
|
XP_016856189.1 |
XM_017000701.1 |
2082 |
Missense Mutation |
CAC,CGC |
H516R |
XP_016856190.1 |
XM_017000702.1 |
2082 |
Missense Mutation |
CAC,CGC |
H504R |
XP_016856191.1 |
XM_017000703.1 |
2082 |
Missense Mutation |
CAC,CGC |
H504R |
XP_016856192.1 |
XM_017000704.1 |
2082 |
Missense Mutation |
CAC,CGC |
H494R |
XP_016856193.1 |
XM_017000705.1 |
2082 |
Missense Mutation |
CAC,CGC |
H459R |
XP_016856194.1 |
XM_017000706.1 |
2082 |
Missense Mutation |
CAC,CGC |
H445R |
XP_016856195.1 |
XM_017000707.1 |
2082 |
Missense Mutation |
CAC,CGC |
H437R |
XP_016856196.1 |
XM_017000708.1 |
2082 |
Missense Mutation |
CAC,CGC |
H436R |
XP_016856197.1 |
XM_017000709.1 |
2082 |
Missense Mutation |
CAC,CGC |
H427R |
XP_016856198.1 |
XM_017000710.1 |
2082 |
Missense Mutation |
CAC,CGC |
H423R |
XP_016856199.1 |
XM_017000711.1 |
2082 |
Missense Mutation |
CAC,CGC |
H414R |
XP_016856200.1 |
XM_017000712.1 |
2082 |
Missense Mutation |
CAC,CGC |
H414R |
XP_016856201.1 |
XM_017000713.1 |
2082 |
Missense Mutation |
CAC,CGC |
H406R |
XP_016856202.1 |
- Gene
- SLFNL1
- Gene Name
- schlafen like 1
There are no transcripts associated with this gene.
- Gene
- SLFNL1-AS1
- Gene Name
- SLFNL1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details