Product Details

SNP ID
rs143627824
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27394353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGATGGCCTGACCACATGTGG[C/T]GTGGTTTATCCACTCTCCAAGAACC
Phenotype
MIM: 600241 MIM: 605875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR3 PubMed Links

Gene Details

Gene
GPR3
Gene Name
G protein-coupled receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005281.3 654 Silent Mutation GGC,GGT G185G NP_005272.1
Gene
WASF2
Gene Name
WAS protein family member 2
There are no transcripts associated with this gene.

View Full Product Details