Product Details

SNP ID
rs143811653
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:48112971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATCCACTGAACACCAACTCCCT[A/G]TCCCCTCACGTGGCTAGCCTGTGGA
Phenotype
MIM: 616392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SKINT1L PubMed Links

Gene Details

Gene
SKINT1L
Gene Name
Skint1 like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details