Product Details

SNP ID
rs143897532
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:233614244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCGCTCGGCCTGGTGCTTCGGC[A/T]TCCTGGTGCTGGGCTACTTGCTCTA
Phenotype
MIM: 601745
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNK1 PubMed Links

Gene Details

Gene
KCNK1
Gene Name
potassium two pore domain channel subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002245.3 241 Missense Mutation ATC,TTC I25F NP_002236.1
XM_011544184.2 241 Intron XP_011542486.1
Gene
MIR4427
Gene Name
microRNA 4427
There are no transcripts associated with this gene.

View Full Product Details