Product Details

SNP ID
rs143947332
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:158563104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGATAATGTTGGGTCCACAGTA[A/G]GACAATTGGGAAATAAAAAGAAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6P1 PubMed Links

Gene Details

Gene
OR6P1
Gene Name
olfactory receptor family 6 subfamily P member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160325.1 501 Silent Mutation TCC,TCT S167S NP_001153797.1

View Full Product Details