Product Details

SNP ID
rs144113895
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:173624748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACAACATTGAACTTAATTTTCAT[A/C]CCTTCTGACATTCAAACCACCTTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD45 PubMed Links

Gene Details

Gene
ANKRD45
Gene Name
ankyrin repeat domain 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198493.2 Intron NP_940895.1
XM_011509466.2 Intron XP_011507768.1
XM_011509467.1 Intron XP_011507769.1
XM_011509468.1 Intron XP_011507770.1
XM_011509470.1 Intron XP_011507772.1
XM_011509471.2 Intron XP_011507773.1
XM_017001123.1 Intron XP_016856612.1
XM_017001124.1 Intron XP_016856613.1
Gene
LOC730159
Gene Name
uncharacterized LOC730159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195190.1 Intron NP_001182119.1
XM_011509968.2 Intron XP_011508270.1

View Full Product Details