Product Details

SNP ID
hCV166519607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:150260157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTATTCTGCCTTGCAGGTCAAC[A/C]CTGGACGTATGAGGGTGAGCAGATC
Phenotype
MIM: 607629 MIM: 604832
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APH1A PubMed Links

Gene Details

Gene
APH1A
Gene Name
aph-1 homolog A, gamma-secretase subunit
There are no transcripts associated with this gene.

Gene
CA14
Gene Name
carbonic anhydrase 14
There are no transcripts associated with this gene.

View Full Product Details