Product Details
- SNP ID
-
rs144388879
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:183630504 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACATAGCACTGCTATTGTCAGAC[A/G]GGCTCTCAAATCCTTTATAAATATA
- Phenotype
-
MIM: 604227
MIM: 605667
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARPC5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2231240] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARPC5
- Gene Name
- actin related protein 2/3 complex subunit 5
- Gene
- RGL1
- Gene Name
- ral guanine nucleotide dissociation stimulator like 1
There are no transcripts associated with this gene.
View Full Product Details