Product Details

SNP ID
rs144414153
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:158563178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCCAAGAGGCAGCAGCAAGGC[A/G]AGTGGCCAGACTGGAAGGCATGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6P1 PubMed Links

Gene Details

Gene
OR6P1
Gene Name
olfactory receptor family 6 subfamily P member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160325.1 427 Missense Mutation CGC,TGC R143C NP_001153797.1

View Full Product Details