Product Details

SNP ID
rs144429005
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:26706696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTTCTCCTGTCTGGGGTAGAC[A/G]TTGTTGCTTTCTGTTGTCCTTAGAG
Phenotype
MIM: 603024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID1A PubMed Links

Gene Details

Gene
ARID1A
Gene Name
AT-rich interaction domain 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006015.4 Intron NP_006006.3
NM_139135.2 Intron NP_624361.1

View Full Product Details