Product Details

SNP ID
rs144787725
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:40260871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATAAAACAACAACTCATGTACC[A/T]CCGGAGTTAGGACAGATCATGGATT
Phenotype
MIM: 606480
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMCO2 PubMed Links

Gene Details

Gene
TMCO2
Gene Name
transmembrane and coiled-coil domains 2
There are no transcripts associated with this gene.

Gene
ZMPSTE24
Gene Name
zinc metallopeptidase STE24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005857.4 378 Silent Mutation CCA,CCT P52P NP_005848.2

View Full Product Details