Product Details

SNP ID
rs144800430
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:1203871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCCACAGGTCTCCCAGCCGCCC[C/G]TTCTCCTCTGCCGATCGCTCGCCCC
Phenotype
MIM: 603905 MIM: 600315
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TNFRSF18 PubMed Links

Gene Details

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004195.2 816 Missense Mutation AAC,AAG N233K NP_004186.1
NM_148901.1 816 Missense Mutation ACG,AGG T163R NP_683699.1
NM_148902.1 816 Missense Mutation AAC,AAG N226K NP_683700.1
XM_017002722.1 816 Missense Mutation ACG,AGG T255R XP_016858211.1
Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
There are no transcripts associated with this gene.

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
There are no transcripts associated with this gene.

View Full Product Details