Product Details

SNP ID
rs145110984
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:159588169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCATGTAAACTTGATCACACCG[C/T]TGGAGAAGCCTCTACAGAACTTTAC
Phenotype
MIM: 104770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APCS PubMed Links

Gene Details

Gene
APCS
Gene Name
amyloid P component, serum
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001639.3 229 Silent Mutation CTG,TTG L45L NP_001630.1

View Full Product Details