Product Details

SNP ID
rs145137014
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:167053488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTACTAGGGCTTCAGGGAAAGC[A/C]AAAGCCATGAGGCCCAGGCCCTGCC
Phenotype
MIM: 602171
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 1978 UTR 3 NP_005805.1
XM_017000005.1 1978 UTR 3 XP_016855494.1

View Full Product Details