Product Details

SNP ID
rs145824319
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:23804501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGCCTCCAAGTCCTGCCACAG[A/G]AGAGTCCACGACACTCACTCCCATC
Phenotype
MIM: 606953 MIM: 613898 MIM: 616143
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALE PubMed Links

Gene Details

Gene
GALE
Gene Name
UDP-galactose-4-epimerase
There are no transcripts associated with this gene.

Gene
HMGCL
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000191.2 606 Missense Mutation CCT,TCT P259S NP_000182.2
NM_001166059.1 606 Missense Mutation CCT,TCT P188S NP_001159531.1
Gene
LYPLA2
Gene Name
lysophospholipase II
There are no transcripts associated with this gene.

View Full Product Details