Product Details

SNP ID
rs146201749
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:156841006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTAGTAGAAGGAGAGGAGGCGG[A/G]AGGAGGGCCGCAGCTCCTCCTGTAT
Phenotype
MIM: 147671 MIM: 191315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INSRR PubMed Links

Gene Details

Gene
INSRR
Gene Name
insulin receptor related receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014215.2 4060 Missense Mutation TCC,TTC S1254F NP_055030.1
Gene
NTRK1
Gene Name
neurotrophic receptor tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007792.1 4060 Intron NP_001007793.1
NM_001012331.1 4060 Intron NP_001012331.1
NM_002529.3 4060 Intron NP_002520.2

View Full Product Details