Product Details
- SNP ID
-
rs146223622
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:20744941 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTGGATGAGGGTCTGGTCTTCTT[C/G]GCAGGCGTTTTGGCCTTAGGAGGTG
- Phenotype
-
MIM: 616072
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HP1BP3
PubMed Links
Gene Details
- Gene
- HP1BP3
- Gene Name
- heterochromatin protein 1 binding protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016287.3 |
2094 |
Silent Mutation |
GCC,GCG |
A506A |
NP_057371.2 |
XM_005245875.4 |
2094 |
Silent Mutation |
GCC,GCG |
A506A |
XP_005245932.1 |
XM_005245876.3 |
2094 |
Silent Mutation |
GCC,GCG |
A506A |
XP_005245933.1 |
XM_005245877.4 |
2094 |
Silent Mutation |
GCC,GCG |
A506A |
XP_005245934.1 |
XM_005245878.4 |
2094 |
Silent Mutation |
GCC,GCG |
A506A |
XP_005245935.1 |
XM_005245879.4 |
2094 |
Silent Mutation |
GCC,GCG |
A468A |
XP_005245936.1 |
XM_011541532.1 |
2094 |
Silent Mutation |
GCC,GCG |
A468A |
XP_011539834.1 |
XM_011541533.1 |
2094 |
Silent Mutation |
GCC,GCG |
A468A |
XP_011539835.1 |
XM_011541534.2 |
2094 |
Silent Mutation |
GCC,GCG |
A468A |
XP_011539836.1 |
XM_017001393.1 |
2094 |
Silent Mutation |
GCC,GCG |
A506A |
XP_016856882.1 |
XM_017001394.1 |
2094 |
Silent Mutation |
GCC,GCG |
A468A |
XP_016856883.1 |
View Full Product Details